Learn More
BioVision IncSupplier Diversity Partner 8-hydroxy-2'-deoxyguanosine (8-OHdG) ELISA Kit: An in vitro enzyme linked immunosorbent assay (ELISA) for the quantitative measurement of 8-OHdG in serum, plasma, tissue homogenates, culture supernatants and other biological fluids. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K4160100
8-hydroxy-2'-deoxyguanosine (8-OHdG) is an oxidized derivative of deoxyguanosine. Concentrations of 8-oxo-dG within a cell are a measurement of oxidative stress. It is considered as a critical biomarker of oxidative stress and carcinogenesis. This ELISA kit uses Competitive-ELISA as the method. The microtiter plate provided in this kit has been pre-coated with 8-OHdG. During the reaction,8-OHdG in the sample or standard competes with a fixed amount of 8-OHdG on the solid phase supporter for sites on the Biotinylated Detection Antibody specific to 8-OHdG. The concentration of 8-OHdG in the samples is determined by comparing the O.D. of the samples to the standard curve. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-113-2283