Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-8-Phenylmenthol, 97%, Acros Organics

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
65253-04-5
C16H24O
232.367
MFCD00010501
WTQIZFCJMGWUGZ-BPLDGKMQSA-N
--8-phenylmenthol,--phenmenthol,1r,2s,5r-5-methyl-2-2-phenylpropan-2-yl cyclohexanol,cyclohexanol, 5-methyl-2-1-methyl-1-phenylethyl-, 1r,2s,5r,1r,2s,5r-5-methyl-2-2-phenylpropan-2-yl cyclohexan-1-ol,1r,2s,5r-5-methyl-2-1-methyl-1-phenylethyl cyclohexanol,cyclohexanol,5-methyl-2-1-methyl-1-phenylethyl-, 1r,2s,5r
2725001
(1R,2S,5R)-5-methyl-2-(2-phenylpropan-2-yl)cyclohexan-1-ol
CC1CCC(C(C1)O)C(C)(C)C2=CC=CC=C2
65253-04-5
C16H24O
232.367
MFCD00010501
WTQIZFCJMGWUGZ-BPLDGKMQSA-N
--8-phenylmenthol,--phenmenthol,1r,2s,5r-5-methyl-2-2-phenylpropan-2-yl cyclohexanol,cyclohexanol, 5-methyl-2-1-methyl-1-phenylethyl-, 1r,2s,5r,1r,2s,5r-5-methyl-2-2-phenylpropan-2-yl cyclohexan-1-ol,1r,2s,5r-5-methyl-2-1-methyl-1-phenylethyl cyclohexanol,cyclohexanol,5-methyl-2-1-methyl-1-phenylethyl-, 1r,2s,5r
2725001
(1R,2S,5R)-5-methyl-2-(2-phenylpropan-2-yl)cyclohexan-1-ol
CC1CCC(C(C1)O)C(C)(C)C2=CC=CC=C2
Infrared Spectrum Authentic
Boiling Point 130.0°C (1.0hPa)
Density 1.0000g/mL
Color Colorless to Yellow
Specific Rotation − 24.00
Flash Point >110°C
Specific Rotation Condition −24.00 (20.00°C c=2,C2H5OH)
Packaging Glass bottle
Refractive Index 1.5285 to 1.5315
Quantity 250mg
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 232.37
Physical Form Viscous Liquid
Percent Purity 97%
Chemical Name or Material (-)-8-Phenylmenthol
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.