Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Quinolinesulfonyl chloride, 98%, Acros Organics™

Click to view available options
Quantity:
5g
25g
18704-37-5
C9H6ClNO2S
227.662
JUYUYCIJACTHMK-UHFFFAOYSA-N
8-quinolinesulfonyl chloride,8-chlorosulfonyl-1-benzazine,8-quinolylsulfonyl chloride,quinoline-8-sulfonic acid chloride,unii-5zik3tjs9m,8-quinolinesulfonylchloride,quinoline-8-sulphonyl chloride,5zik3tjs9m,pubchem5873,acmc-1busp
29220
quinoline-8-sulfonyl chloride
C1=CC2=C(C(=C1)S(=O)(=O)Cl)N=CC=C2
18704-37-5
C9H6ClNO2S
227.662
JUYUYCIJACTHMK-UHFFFAOYSA-N
8-quinolinesulfonyl chloride,8-chlorosulfonyl-1-benzazine,8-quinolylsulfonyl chloride,quinoline-8-sulfonic acid chloride,unii-5zik3tjs9m,8-quinolinesulfonylchloride,quinoline-8-sulphonyl chloride,5zik3tjs9m,pubchem5873,acmc-1busp
29220
quinoline-8-sulfonyl chloride
C1=CC2=C(C(=C1)S(=O)(=O)Cl)N=CC=C2
Quantity 25g
Boiling Point 306.0°C
Infrared Spectrum Authentic
Melting Point 126.0°C to 130.0°C
Solubility Information Solubility in water: reacts.
Formula Weight 227.67
Percent Purity 98%
Chemical Name or Material 8-Quinolinesulfonyl chloride

GHS H Statement Causes severe skin burns and eye damage.

GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

Danger