Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diversified Spaces™ 8-Station Service Center with Sink
Double-faced table that provides a fully equipped science center for eight students. Diversified Spaces™™ 8-Station Service Center contains a solid epoxy resin sink with cold water faucets for every two students.
Supplier: Diversified Spaces™ C2424K
Description
- Eight student double faced workstation that provides 2 sinks, four multiservice fixtures, four GFI protected AC duplex receptacles and four rod sockets
- Includes eight book compartments in the aprons
- Constructed of solid oak and oak hardwood veneers
- The unit is finished with a chemical resistant, earth-friendly UV finish, rubber base molding and leg boots
- Diversified Spaces™ is SEFA compliant and MAS Certified Green
- Limited Lifetime Warranty
- Made in the U.S.A.
Specifications
48 x 132 x 36 in. | |
8 | |
Chemical Resistant, Earth-friendly UV | |
Oak with Oak Veneer |
0.75 in. phenolic resin surface | |
SEFA compliant and MAS Certified Green | |
2 sinks; 4 multiservice fixtures; 4 GFI protected AC duplex receptacles; 4 rod sockets; 4 locking drawers and locking cupboard w/ shelf |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction