Learn More
Diversified Spaces™ 8-Station Service Center with Sink
SCI_ED
Double-faced table that provides a fully equipped science center for eight students. Diversified Spaces™™ 8-Station Service Center contains a solid epoxy resin sink with cold water faucets for every two students.
Supplier:  Diversified Spaces™ P7182BK34L
 View more versions of this product
Catalog No. S38134
Add to Cart
Description
Description
  • Eight student double faced workstation that provides 2 sinks, four multiservice fixtures, four GFI protected AC duplex receptacles and four rod sockets
  • Includes eight book compartments in the aprons
  • Constructed of solid oak and oak hardwood veneers
  • The unit is finished with a chemical resistant, earth-friendly UV finish, rubber base molding and leg boots
  • Diversified Spaces™ is SEFA compliant and MAS Certified Green
  • Limited Lifetime Warranty
  • Made in the U.S.A.
Specifications
Specifications
SEFA compliant and MAS Certified Green
1 in. Solid epoxy resin surface
48 x 132 x 36 in.
Oak, Hardwood and Oak Veneer
8
2 sinks; 4 multiservice fixtures; 4 GFI protected AC duplex receptacles; 4 rod sockets and four locking drawers and a locking cupboard with shelf
Chemical Resistant, Earth-friendly UV