Alfa Aesar™ 8-(Tri-n-butylstannyl)quinoline, 95%

Manufacturer: Alfa Aesar™ H5147103

Quantity 1g

 View more versions of this product

Catalog No. AAH5147103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 478282-21-2
    Molecular Formula C21H33NSn
    Formula Weight 418.19
    MDL Number MFCD09265482
    UN Number UN2788
    Synonym 8-Quinolinetri-n-butylstannane; (Tri-n-butyl)-8-quinolinyltin
    Chemical Name or Material 8-(Tri-n-butylstannyl)quinoline

    Specifications

    Formula Weight 418.19
    Molecular Formula C21H33NSn
    MDL Number MFCD09265482
    CAS 478282-21-2
    UN Number UN2788
    Synonym 8-Quinolinetri-n-butylstannane; (Tri-n-butyl)-8-quinolinyltin
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H372-H312-H315-H319
    Toxic if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280-P273-P309-P310-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
    Quantity 1g
    Chemical Name or Material 8-(Tri-n-butylstannyl)quinoline
    Assay 95%

    GHS H Statement
    H301-H372-H312-H315-H319
    Toxic if swallowed.
    Causes damage to organs through prolonged or repeated exposure.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P273-P309-P310-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit