Learn More
Greiner Bio-One 8-Well Polystyrene Round Bottom Strip Plates
Free of detectable DNase/RNase, human DNA and pyrogens.
$250.00
Specifications
Sterility Non-sterile
Bottom U
Format Strip Plate
Material Polystyrene
No. per Case 400
View More Specs
Products 1
Catalog Number Mfr. No. Surface Treatment Price Quantity & Availability  
Catalog Number Mfr. No. Surface Treatment Price Quantity & Availability  
07-000-849
View Documents
Greiner Bio-One
767001
Microlon 200
Case of 400 for $250.00
Add to cart
Description
Description
  • 1 x 8 strips
  • Non-Sterile
  • Round Bottom
Specifications
Specifications
Non-sterile
Strip Plate
400
U
Polystyrene
8