Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCS 8200 Series 3-Layer Vinyl Floor Kit
Provides reliable path to ground and eliminates static charge generation as a worksurface table mat, shelf liner, and cart liner
Supplier: SCS 8211
Description
- 3-Layer Vinyl Material
Provides reliable path to ground and eliminates static charge generation as a worksurface table mat, shelf liner, and cart liner. - Dissipative (1 x 106 < 1 x 109 ohms Rtt) Top Color Layer
Meets ANSI/ESD S20.20 worksurface required limit and recommendation of ANSI/ESD S4.1 for contact with ESD susceptible items. Electricals will not drift for the life of the mat. - Highly, Conductive Scrim Middle Layer
Provides main discharge path to ground. - Dissipative (1 x 106 < 1 x 109 ohms Rtt) Foam Bottom Layer
Provides durable, nonskid cushion, choice for components susceptible to physical shock. - Compatible with Constant Monitors
- Lead-free RoHS 2, REACH and Conflict Minerals compliant
- Made in the United States of America
Specifications
121.92 cm | |
Vinyl | |
24 in. | |
24 x 48 in. |
Brown | |
48 in. | |
60.96 cm | |
Wrist Strap Grounding System and Snap Fasteners (snaps installed on long side of mat) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction