Learn More
SCS 8200 Series 3-Layer Vinyl Floor Kit
Provides reliable path to ground and eliminates static charge generation as a worksurface table mat, shelf liner, and cart liner
Supplier:  SCS 8214
 View more versions of this product
Catalog No. 18-688-312
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • 3-Layer Vinyl Material
    Provides reliable path to ground and eliminates static charge generation as a worksurface table mat, shelf liner, and cart liner.
  • Dissipative (1 x 106 < 1 x 109 ohms Rtt) Top Color Layer
    Meets ANSI/ESD S20.20 worksurface required limit and recommendation of ANSI/ESD S4.1 for contact with ESD susceptible items. Electricals will not drift for the life of the mat.
  • Highly, Conductive Scrim Middle Layer
    Provides main discharge path to ground.
  • Dissipative (1 x 106 < 1 x 109 ohms Rtt) Foam Bottom Layer
    Provides durable, nonskid cushion, choice for components susceptible to physical shock.
  • Compatible with Constant Monitors
  • Lead-free RoHS 2, REACH and Conflict Minerals compliant
  • Made in the United States of America
Specifications
Specifications
24 x 48 in.
121.92 cm
24 in.
Vinyl
Wrist Strap Grounding System and Snap Fasteners (snaps installed on long side of mat)
48 in.
60.96 cm
Blue
Product Suggestions
Product Suggestions