Learn More
SCS 8200 Series 3-Layer Vinyl Floor Kit
Provides reliable path to ground and eliminates static charge generation as a worksurface table mat, shelf liner, and cart liner
$90.10 - $1335.00
Catalog Number | Mfr. No. | Dimensions (L x W) | Color | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Color | Includes | Price | Quantity & Availability | |||
18-688-311
|
SCS
8213 |
24 x 48 in. | Gray | Wrist Strap Grounding System and Snap Fasteners (snaps installed on long side of mat) |
Each for $91.90
|
|
18-688-312
|
SCS
8214 |
24 x 48 in. | Blue | Wrist Strap Grounding System and Snap Fasteners (snaps installed on long side of mat) |
Each for $91.90
|
|
18-688-310
|
SCS
8211 |
24 x 48 in. | Brown | Wrist Strap Grounding System and Snap Fasteners (snaps installed on long side of mat) |
Each for $90.10
|
|
18-688-308
|
SCS
8203 |
48 x 72 in. | Gray | 15 ft. (4.6 m) Ground Cord and 2 Snap Fasteners |
Each for $226.50
|
|
18-688-309
|
SCS
8204 |
48 x 72 in. | Blue | 15 ft. (4.6 m) Ground Cord and 2 Snap Fasteners |
Each for $226.50
|
|
18-688-318
|
SCS
8263 |
24 ft. x 24 in. | Gray | Grounding Hardware not included |
Each for $351.50
|
|
18-688-319
|
SCS
8264 |
24 ft. x 24 in. | Blue | Grounding Hardware not included |
Each for $351.50
|
|
18-688-317
|
SCS
8253 |
24 ft. x 48 in. | Gray | Grounding Hardware not included |
Each for $706.00
|
|
18-688-314
|
SCS
8233 |
50 ft. x 24 in. | Gray | Grounding Hardware not included |
Each for $667.50
|
|
18-688-315
|
SCS
8234 |
50 ft. x 24 in. | Blue | Grounding Hardware not included |
Each for $667.00
|
|
18-688-313
|
SCS
8231 |
50 ft. x 24 in. | Brown | Grounding Hardware not included |
Each for $667.50
|
|
18-688-316
|
SCS
8241 |
50 ft. x 30 in. | Brown | Grounding Hardware not included |
Each for $1,069.00
|
|
18-688-320
|
SCS
8274 |
50 ft. x 36 in. | Blue | Grounding Hardware not included |
Each for $1,215.00
|
|
18-688-322
|
SCS
82A3 |
50 ft. x 48 in. | Gray | Grounding Hardware not included |
Each for $1,335.00
|
|
18-688-323
|
SCS
82A4 |
50 ft. x 48 in. | Blue | Grounding Hardware not included |
Each for $1,335.00
|
|
18-688-321
|
SCS
82A1 |
50 ft. x 48 in. | Brown | Grounding Hardware not included |
Each for $1,335.00
|
|
Description
- 3-Layer Vinyl Material
Provides reliable path to ground and eliminates static charge generation as a worksurface table mat, shelf liner, and cart liner. - Dissipative (1 x 106 < 1 x 109 ohms Rtt) Top Color Layer
Meets ANSI/ESD S20.20 worksurface required limit and recommendation of ANSI/ESD S4.1 for contact with ESD susceptible items. Electricals will not drift for the life of the mat. - Highly, Conductive Scrim Middle Layer
Provides main discharge path to ground. - Dissipative (1 x 106 < 1 x 109 ohms Rtt) Foam Bottom Layer
Provides durable, nonskid cushion, choice for components susceptible to physical shock. - Compatible with Constant Monitors
- Lead-free RoHS 2, REACH and Conflict Minerals compliant
- Made in the United States of America
Specifications
Vinyl |