Learn More
Cell Biolabs IncSupplier Diversity Partner 8ISOPROSTAGLANDINF2A ELISA 32W  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner STA337T
The OxiSelect 8-iso-Prostaglandin F2a ELISA (Trial Size) measures 8-isoprostane in urine, plasma, serum, or cell lysates. Standard included against which unknown samples are measured. 32 wells including standards and unknown samples.
Catalog No. 50-673-529
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.