Thermo Scientific™ 8mm Assembled Clear Autosampler Vial Kits Available on GSA/VA Contract for Federal Government customers only.
Convenience kits save time and reduce the risk of contamination. Each convenience kit includes 100 vials pre-assembled with 100 screw thread caps and septa.
Manufacturer: Thermo Scientific™ C401357
 View more versions of this product
Catalog No. 03-375-50E
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Septa Red PTFE/White Silicone
    Type Assembled Vial Kit
    Vial Volume (Metric) 2mL
    Description Clear Glass, flanged; PP Black Cap; Red PTFE/White Silicone septum; in storage tray
    Material Glass
    Color Clear
    Patched No
    No. per Pack 100
    Vial Closure Size 8-425mm
    Closure Color Black
    Closure Material Polypropylene
    Closure Type Polypropylene, Flanged
    Item Description Assembled Kit
    Save time during sample preparation and reduce the risk of contamination by using Thermo Scientific™ 8mm Clear Vial Convenience Kits. Each kit includes 100 clear glass vials pre-assembled with 100 screw thread caps and septa. They are packaged in convenient vial trays with clear covers or in economical polybags. Choose from the Thermo Scientific™, Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ brands.
    • Packaged in convenient vial trays with clear covers or in economical polybags
    • Vial trays are stackable for easy storage
    • All caps are manufactured from virgin polypropylene unless noted
    Includes: 
    100 vials with pre-attached caps and septa
  • Description & Specifications
Related Products
Product Content Feedback