Learn More
Thermo Scientific™ 8mm Autosampler Vial Screw Thread Caps Available on GSA/VA Contract for Federal Government customers only.
Available in an variety of styles to match your application. Pre-assembled caps and septa minimize contamination.
Manufacturer:  Thermo Scientific™ 60180665
 View more versions of this product
Catalog No. 03-246-305
  Check Availability
Add to cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Thermo Scientific? 8mm Autosampler Vial Screw Thread Caps feature an 8-425 thread finish and are available in an assortment of styles to match your HPLC, GC or IC application needs. Convenient pre-assembled caps and septa minimize contamination from handling. Choose from Thermo Scientific?, Thermo Scientific? National?, and Thermo Scientific? Chromacol? brands.
  • Pre-assembled caps and septa are convenient to minimize contamination from handling
  • Fully compatible with all Target DP vials
  • Easy-on, easy-off convenience with just half-a-turn
  • Bonded caps feature a PTFE/Silicone septum electrolytically bonded into the cap, reducing septa push-though when used with blunt tip needles on high-speed autosamplers
Description & Specifications
Specifications
Pink
Ivory PTFE/Red Rubber
1.0mm
Open Top,Screw Cap
2mL
Polypropylene
8mm
100
8mm Open Top Screw Cap, 8-425 thread, 5.5mm hole