Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Qorpak™ 8 oz. (240mL) Clear Type I Borosilicate Glass Straight Sided Jar with 70-400 Neck Finish
One of our most popular jars is now available in Borosilicate Glass - suggested for critical applications where thermal stability, chemical corrosion resistance and durability are required
Manufacturer:  Qorpak™ 271012
 View more versions of this product
Catalog No. 02-913-104
Description
Description
One of our most popular jars is now available in Borosilicate Glass - suggested for critical applications where thermal stability, chemical corrosion resistance and durability are required. These jars are designed without shoulders for maximum storage capacity and content visibility. Borosilicate glass jars are more resistant to temperature change than regular glass and give greater assurance of your content's integrity. Quickly identify our borosilicate jars from regular jars by the Qorpak™ logo on the bottom of the jar.
Specifications
Specifications
8oz
Straight Sided Round Jar
70-400
240 mL
Borosilicate Glass
Green Thermoset F217 & PTFE Lined Cap Attached