Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Dimethylanthracene, 97%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
781-43-1
C16H14
206.29
MFCD00001262
JTGMTYWYUZDRBK-UHFFFAOYSA-N
anthracene, 9,10-dimethyl,unii-di986077vr,ccris 2742,9,10-dimethyl-anthracene,gnf-pf-2329,acmc-209pdj,9,10-dimethylanthracen,9:10-dimethylanthracene,anthracene,10-dimethyl,9,10-dimethylanthracene
13076
9,10-dimethylanthracene
CC1=C2C=CC=CC2=C(C3=CC=CC=C13)C
781-43-1
C16H14
206.29
MFCD00001262
JTGMTYWYUZDRBK-UHFFFAOYSA-N
anthracene, 9,10-dimethyl,unii-di986077vr,ccris 2742,9,10-dimethyl-anthracene,gnf-pf-2329,acmc-209pdj,9,10-dimethylanthracen,9:10-dimethylanthracene,anthracene,10-dimethyl,9,10-dimethylanthracene
13076
9,10-dimethylanthracene
CC1=C2C=CC=CC2=C(C3=CC=CC=C13)C
Melting Point 180°C to 185°C
Color Yellow
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 05,II,593
Formula Weight 206.29
Physical Form Crystals or Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 9, 10-Dimethylanthracene, 97%
Show More Show Less

EINECSNumber : 212-308-4

RTECSNumber : CA9685000