Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Dimethylanthracene, 97%, Thermo Scientific Chemicals

Catalog Number ac115972500
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
781-43-1
C16H14
206.29
MFCD00001262
JTGMTYWYUZDRBK-UHFFFAOYSA-N
anthracene, 9,10-dimethyl, unii-di986077vr, ccris 2742, 9,10-dimethyl-anthracene, gnf-pf-2329, acmc-209pdj, 9,10-dimethylanthracen, 9:10-dimethylanthracene, anthracene,10-dimethyl, 9,10-dimethylanthracene
13076
9,10-dimethylanthracene
CC1=C2C=CC=CC2=C(C)C2=CC=CC=C12
781-43-1
C16H14
206.29
MFCD00001262
JTGMTYWYUZDRBK-UHFFFAOYSA-N
anthracene, 9,10-dimethyl, unii-di986077vr, ccris 2742, 9,10-dimethyl-anthracene, gnf-pf-2329, acmc-209pdj, 9,10-dimethylanthracen, 9:10-dimethylanthracene, anthracene,10-dimethyl, 9,10-dimethylanthracene
13076
9,10-dimethylanthracene
CC1=C2C=CC=CC2=C(C)C2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 180°C to 185°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 250 mg
Beilstein 05,II,593
Formula Weight 206.29
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 9, 10-Dimethylanthracene, 97%
Show More Show Less
EINECSNumber 212-308-4
RTECSNumber CA9685000

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit