Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 9,10-Dimethylanthracene, 99%

Click to view available options
Quantity:
1g
5g
781-43-1
C16H14
206.288
MFCD00001262
JTGMTYWYUZDRBK-UHFFFAOYSA-N
anthracene, 9,10-dimethyl,unii-di986077vr,ccris 2742,9,10-dimethyl-anthracene,gnf-pf-2329,acmc-209pdj,9,10-dimethylanthracen,9:10-dimethylanthracene,anthracene,10-dimethyl,9,10-dimethylanthracene
13076
9,10-dimethylanthracene
CC1=C2C=CC=CC2=C(C3=CC=CC=C13)C
781-43-1
C16H14
206.288
MFCD00001262
JTGMTYWYUZDRBK-UHFFFAOYSA-N
anthracene, 9,10-dimethyl,unii-di986077vr,ccris 2742,9,10-dimethyl-anthracene,gnf-pf-2329,acmc-209pdj,9,10-dimethylanthracen,9:10-dimethylanthracene,anthracene,10-dimethyl,9,10-dimethylanthracene
13076
9,10-dimethylanthracene
CC1=C2C=CC=CC2=C(C3=CC=CC=C13)C
Applications
9,10-Dimethylanthracene is used as an antioxidant-photosensitizer dual-loaded polymeric micelles with controllable production of reactive oxygen species. It is also used as chemical rate constant actinometer in singlet molecular oxygen reactions.

Solubility
Soluble in toluene. Insoluble in water.

Notes
Incompatible with oxidizing agents.
Melting Point 182°C to 184°C
Quantity 1g
Beilstein 1909028
Formula Weight 206.29
Percent Purity 99%
Chemical Name or Material 9,10-Dimethylanthracene

GHS H Statement
H341-H351
Suspected of causing genetic defects.
Suspected of causing cancer.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CA9685000