Learn More
9,10-Phenanthrenequinone, 95%, Thermo Scientific Chemicals
Description
9,10-Phenanthrenequinon may be used for high quality passivation on silicon (100) surfaces. Quinones may serve as substrates for a variety of flavoenzymes, useful fluorescent compound and quinone. Also used in dyes, as a hardener for dental restoration, for the determination of copper by spectrophotometric method, and for organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
9,10-Phenanthrenequinon may be used for high quality passivation on silicon (100) surfaces. Quinones may serve as substrates for a variety of flavoenzymes, useful fluorescent compound and quinone. Also used in dyes, as a hardener for dental restoration, for the determination of copper by spectrophotometric method, and for organic synthesis.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Melting Point | 207°C to 212°C |
Density | 1.405 |
Boiling Point | >360°C |
Flash Point | 245°C (473°F) |
Quantity | 100 g |
UN Number | UN3077 |
Beilstein | 608838 |
Merck Index | 14,7213 |
Solubility Information | Insoluble in water. |
Formula Weight | 208.22 |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
EINECSNumber | 201-515-5 |
RTECSNumber | SF7875000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.