Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 9(10H)-Acridone, 99%

Click to view available options
Quantity:
1g
5g
25g
578-95-0
C13H9NO
195.221
MFCD00005019
FZEYVTFCMJSGMP-UHFFFAOYSA-N
acridone,9 10h-acridone,acridin-9 10h-one,9-acridone,9 10h-acridinone,acridanone,9-acridanone,acridin-9-one,9-acridinol,9,10-dihydro-9-oxoacridine
2015
CHEBI:50756
10H-acridin-9-one
C1=CC=C2C(=C1)C(=O)C3=CC=CC=C3N2
578-95-0
C13H9NO
195.221
MFCD00005019
FZEYVTFCMJSGMP-UHFFFAOYSA-N
acridone,9 10h-acridone,acridin-9 10h-one,9-acridone,9 10h-acridinone,acridanone,9-acridanone,acridin-9-one,9-acridinol,9,10-dihydro-9-oxoacridine
2015
CHEBI:50756
10H-acridin-9-one
C1=CC=C2C(=C1)C(=O)C3=CC=CC=C3N2
Applications
9(10H)-Acridone is used in the preparation of methyl 9,10-dihydro-9-oxoacridine-10-pentanoate, use for dyeing the organic materials.

Solubility
Insoluble in benzene, chloroform, ether, water and ethanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Melting Point 360°C to 362°C
Quantity 1g
Beilstein 7104
Formula Weight 195.22
Percent Purity 99%
Chemical Name or Material 9(10H)-Acridone

RTECSNumber : AR6976000

TSCA : TSCA