Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-(2-Hydroxyethyl)adenine 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
707-99-3
C7H9N5O
179.183
MFCD04220770
VAQOTZQDXZDBJK-UHFFFAOYSA-N
6-Amino-9-(2-hydroxyethyl)purine
242652
2-(6-aminopurin-9-yl)ethanol
C1=NC2=C(C(=N1)N)N=CN2CCO
707-99-3
C7H9N5O
179.183
MFCD04220770
VAQOTZQDXZDBJK-UHFFFAOYSA-N
6-Amino-9-(2-hydroxyethyl)purine
242652
2-(6-aminopurin-9-yl)ethanol
C1=NC2=C(C(=N1)N)N=CN2CCO
Color White-Yellow
Quantity 25g
Melting Point 242°C
Formula Weight 179.18
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material 9-(2-Hydroxyethyl)adenine