Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
161265-03-8
C39H32OP2
578.63
MFCD00233866
CXNIUSPIQKWYAI-UHFFFAOYSA-N
xantphos,4,5-bis diphenylphosphino-9,9-dimethylxanthene,9,9-dimethyl-4,5-bis diphenylphosphino xanthene,9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphine,dimethylbisdiphenylphosphinoxanthene,unii-nmu72mog9b,nmu72mog9b,9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphane,5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl-diphenylphosphane,4,5-bis-diphenylphosphanyl-9,9-dimethyl-9h-xanthene
636044
(5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl)-diphenylphosphane
CC1(C2=C(C(=CC=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)OC5=C1C=CC=C5P(C6=CC=CC=C6)C7=CC=CC=C7)C
161265-03-8
C39H32OP2
578.63
MFCD00233866
CXNIUSPIQKWYAI-UHFFFAOYSA-N
xantphos,4,5-bis diphenylphosphino-9,9-dimethylxanthene,9,9-dimethyl-4,5-bis diphenylphosphino xanthene,9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphine,dimethylbisdiphenylphosphinoxanthene,unii-nmu72mog9b,nmu72mog9b,9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphane,5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl-diphenylphosphane,4,5-bis-diphenylphosphanyl-9,9-dimethyl-9h-xanthene
636044
(5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl)-diphenylphosphane
CC1(C2=C(C(=CC=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)OC5=C1C=CC=C5P(C6=CC=CC=C6)C7=CC=CC=C7)C
Melting Point 226.0°C to 230.0°C
Color White to Yellow
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Merck Index 15, 10268
Solubility Information Solubility in water: insoluble
Formula Weight 578.63
Physical Form Crystals
Percent Purity 98%
Chemical Name or Material 9, 9-Dimethyl-4, 5-bis(diphenylphosphino)xanthene
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning