Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 9,9-Dimethylfluorene-2-boronic acid, 98%

Click to view available options
Quantity:
250mg
1g
5g
333432-28-3
C15H15BO2
238.093
MFCD08704227
DMDPAJOXRYGXCB-UHFFFAOYSA-N
9,9-dimethyl-9h-fluoren-2-yl boronic acid,9,9-dimethyl-9h-fluoren-2-ylboronic acid,9,9-dimethyl-9h-fluoren-2-yl-boronic acid,9,9-dimethylfluoren-2-boronic acid,9,9-dimethyl-2-fluoreneboronic acid,9,9-dimethyl-9h-fluoren-2-yl-2-boronic acid,9,9-dimethylfluorene-2-boronic acid,9,9-dimethyl-9h-fluoren-2-yl boronic acid,9,9-dimethyl-9h-fluorene-2-ylboronic acid,9,9-dimethyl-9h-fluoren-2-ylboronicacid
22348454
(9,9-dimethylfluoren-2-yl)boronic acid
B(C1=CC2=C(C=C1)C3=CC=CC=C3C2(C)C)(O)O
333432-28-3
C15H15BO2
238.093
MFCD08704227
DMDPAJOXRYGXCB-UHFFFAOYSA-N
9,9-dimethyl-9h-fluoren-2-yl boronic acid,9,9-dimethyl-9h-fluoren-2-ylboronic acid,9,9-dimethyl-9h-fluoren-2-yl-boronic acid,9,9-dimethylfluoren-2-boronic acid,9,9-dimethyl-2-fluoreneboronic acid,9,9-dimethyl-9h-fluoren-2-yl-2-boronic acid,9,9-dimethylfluorene-2-boronic acid,9,9-dimethyl-9h-fluoren-2-yl boronic acid,9,9-dimethyl-9h-fluorene-2-ylboronic acid,9,9-dimethyl-9h-fluoren-2-ylboronicacid
22348454
(9,9-dimethylfluoren-2-yl)boronic acid
B(C1=CC2=C(C=C1)C3=CC=CC=C3C2(C)C)(O)O
Quantity 5g
Beilstein 9563230
Formula Weight 238.09
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 9,9-Dimethylfluorene-2-boronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C