Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 9,9'-Dixanthylidene, 98%

Click to view available options
Quantity:
1g
5g
517-45-3
C26H16O2
360.412
MFCD00597968
SXXWAWNPJCEOGD-UHFFFAOYSA-N
9,9'-dixanthylidene,9-9h-xanthen-9-ylidene-9h-xanthene,dixanthylene,9h-xanthene, 9-9h-xanthen-9-ylidene,9,9'-bixanthenylidene,bixanthylidene,dixanthylidene,.delta.9,9'-bixanthene,delta9,9'-bi 9h-xanthene,sxxwawnpjceogd-uhfffaoysa
68202
9-xanthen-9-ylidenexanthene
C1=CC=C2C(=C1)C(=C3C4=CC=CC=C4OC5=CC=CC=C53)C6=CC=CC=C6O2
517-45-3
C26H16O2
360.412
MFCD00597968
SXXWAWNPJCEOGD-UHFFFAOYSA-N
9,9'-dixanthylidene,9-9h-xanthen-9-ylidene-9h-xanthene,dixanthylene,9h-xanthene, 9-9h-xanthen-9-ylidene,9,9'-bixanthenylidene,bixanthylidene,dixanthylidene,.delta.9,9'-bixanthene,delta9,9'-bi 9h-xanthene,sxxwawnpjceogd-uhfffaoysa
68202
9-xanthen-9-ylidenexanthene
C1=CC=C2C(=C1)C(=C3C4=CC=CC=C4OC5=CC=CC=C53)C6=CC=CC=C6O2
Quantity 1g
Melting Point 318°C to 320°C
Formula Weight 360.41
Percent Purity 98%
Chemical Name or Material 9,9'-Dixanthylidene