Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 9,9'-Spirobifluorene-2-boronic acid, 98%

Click to view available options
Quantity:
250mg
1g
5g
236389-21-2
C25H17BO2
360.219
MFCD13248568
WDDLHUWVLROJLA-UHFFFAOYSA-N
9,9'-spirobi fluoren-2-ylboronic acid,9,9'-spirobi 9h-fluorene-2-boronic acid,b-9,9'-spirobi 9h-fluoren-2'-yl-boronic acid,9,9'-spirobi fluorene-2-ylboronic acid,boronic acid, b-9,9'-spirobi 9h-fluoren-2'-yl,9,9'-spirobi 9h-fluoren-2'-yl-boronic acid,boronic acid, 9,9'-spirobi 9h-fluoren-2-yl,acmc-209g4k,9,9'-spirobifluorene-2-boronic acid,9,9'-spirobi fluorene-2-yl boronic acid
22564369
9,9'-spirobi[fluorene]-2-ylboronic acid
B(C1=CC2=C(C=C1)C3=CC=CC=C3C24C5=CC=CC=C5C6=CC=CC=C46)(O)O
236389-21-2
C25H17BO2
360.219
MFCD13248568
WDDLHUWVLROJLA-UHFFFAOYSA-N
9,9'-spirobi fluoren-2-ylboronic acid,9,9'-spirobi 9h-fluorene-2-boronic acid,b-9,9'-spirobi 9h-fluoren-2'-yl-boronic acid,9,9'-spirobi fluorene-2-ylboronic acid,boronic acid, b-9,9'-spirobi 9h-fluoren-2'-yl,9,9'-spirobi 9h-fluoren-2'-yl-boronic acid,boronic acid, 9,9'-spirobi 9h-fluoren-2-yl,acmc-209g4k,9,9'-spirobifluorene-2-boronic acid,9,9'-spirobi fluorene-2-yl boronic acid
22564369
9,9'-spirobi[fluorene]-2-ylboronic acid
B(C1=CC2=C(C=C1)C3=CC=CC=C3C24C5=CC=CC=C5C6=CC=CC=C46)(O)O
Color White
Quantity 1g
Beilstein 10478712
Formula Weight 360.22
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 9,9'-Spirobifluorene-2-boronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning