Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Amino-1,2,3,4-tetrahydroacridine Hydrochloride Hydrate 95%, ACROS Organics™

Click to view available options
Quantity:
1g
321-64-2
C13H14N2
198.269
MFCD00150067
YLJREFDVOIBQDA-UHFFFAOYSA-N
tacrine,tetrahydroaminacrine,tetrahydroaminoacridine,1,2,3,4-tetrahydro-9-acridinamine,cognex,tetrahydroaminocrin,tetrahydroaminocrine,9-amino-1,2,3,4-tetrahydroacridine,romotal,tacrinum
1935
CHEBI:45980
1,2,3,4-tetrahydroacridin-9-amine
C1CCC2=NC3=CC=CC=C3C(=C2C1)N
321-64-2
C13H14N2
198.269
MFCD00150067
YLJREFDVOIBQDA-UHFFFAOYSA-N
tacrine,tetrahydroaminacrine,tetrahydroaminoacridine,1,2,3,4-tetrahydro-9-acridinamine,cognex,tetrahydroaminocrin,tetrahydroaminocrine,9-amino-1,2,3,4-tetrahydroacridine,romotal,tacrinum
1935
CHEBI:45980
1,2,3,4-tetrahydroacridin-9-amine
C1CCC2=NC3=CC=CC=C3C(=C2C1)N
Water 14% max. (K.F.)
Infrared Spectrum Authentic
Quantity 1g
Color Yellow
Assay Percent Range 98%
Formula Weight 234.72
Physical Form Crystalline Powder
Percent Purity ≥97.5% (on dry substance)
Chemical Name or Material 9-Amino-1,2,3,4-tetrahydroacridine hydrochloride hydrate