Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 9-Aminoacridine hydrochloride hydrate, 99%

Manufacturer:  Alfa Aesar™ B2435614

Quantity 25g

 View more versions of this product

Catalog No. AAB2435614


  Check Availability
Add to cart

Specifications

Description

Applications
9-Aminoacridine hydrochloride is a known mutagen and DNA modifier.

Solubility
Soluble in water (3.3 g/L) at 20°C.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

52417-22-8
230.69 (Anhydrous)
UN2811
9-Aminoacridine hydrochloride hydrate
C13H10N2·HCl·xH2O
MFCD00150071
9-Acridinamine hydrochloride hydrate

Specifications

52417-22-8
230.69 (Anhydrous)
UN2811
9-Acridinamine hydrochloride hydrate
9-Aminoacridine hydrochloride hydrate
C13H10N2·HCl·xH2O
MFCD00150071
>300°C
25g
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H341
Toxic if swallowed.
Suspected of causing genetic defects.

GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : AR7350000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit