Alfa Aesar™ 9-Aminoacridine hydrochloride hydrate, 99%

Manufacturer: Alfa Aesar™ B2435606

Quantity 5g

 View more versions of this product

Catalog No. AAB2435606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 52417-22-8
    Molecular Formula C13H10N2·HCl·xH2O
    Formula Weight 230.69 (Anhydrous)
    MDL Number MFCD00150071
    UN Number UN2811
    Synonym 9-Acridinamine hydrochloride hydrate
    Chemical Name or Material 9-Aminoacridine hydrochloride hydrate

    Specifications

    Formula Weight 230.69 (Anhydrous)
    Molecular Formula C13H10N2·HCl·xH2O
    MDL Number MFCD00150071
    UN Number UN2811
    Melting Point >300°C
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H301-H341
    Toxic if swallowed.
    Suspected of causing genetic defects.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    RTECS Number AR7350000
    TSCA TSCA
    CAS 52417-22-8
    Synonym 9-Acridinamine hydrochloride hydrate
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P280h-P309-P310
    Wear protective gloves/clothing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Chemical Name or Material 9-Aminoacridine hydrochloride hydrate
    Assay 99%

    Description

    Applications
    9-Aminoacridine hydrochloride is a known mutagen and DNA modifier.

    Solubility
    Soluble in water (3.3 g/L) at 20°C.

    Notes
    Store at room temperature. Incompatible with oxidizing agents.

    GHS H Statement
    H301-H341
    Toxic if swallowed.
    Suspected of causing genetic defects.

    GHS P Statement
    P280h-P309-P310
    Wear protective gloves/clothing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit