Alfa Aesar™ 9-Aminophenanthrene, 96%

Manufacturer: Alfa Aesar™ H5577303

Quantity 1g

 View more versions of this product

Catalog No. AAH5577303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 947-73-9
    Formula Weight 193.24
    MDL Number MFCD00001177
    Synonym 9-Phenanthrenamine
    Chemical Name or Material 9-Aminophenanthrene

    Specifications

    Formula Weight 193.24
    Melting Point 137° to 139°C
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Color White to brown
    Physical Form Solid
    MDL Number MFCD00001177
    CAS 947-73-9
    Health Hazard 1 Warning
    Quantity 1g
    Synonym 9-Phenanthrenamine
    RTECS Number SG0175000
    Chemical Name or Material 9-Aminophenanthrene
    Assay 96%

    Description

    Applications
    9-Aminophenanthrene can be used as a model ligand to study the binding of 9-aminophenanthrene to individual subsites within the active site of CYP(eryF). It is also used as a fluorescent-labeling reagent for free fatty acids (FFA),examination through HPLC.

    Solubility
    It is slightly soluble in water 0.00581 mg/mL.

    Notes
    Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit