Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Anthracenemethanol, 97%, Thermo Scientific Chemicals

Catalog Number aaa1518809
Click to view available options
Quantity:
10 g
50 g
250 g
1468-95-7
C15H12O
208.26
MFCD00001264
JCJNNHDZTLRSGN-UHFFFAOYSA-N
9-anthracenemethanol, 9-hydroxymethylanthracene, 9-anthrylcarbinol, 9-methylolanthracene, 9-anthrylmethanol, 9-hydroxymethylantracene, 9-anthracene methanol, 9-hydroxymethyl anthracene, anthracene-9-carbinol, ccris 7297
73848
anthracen-9-ylmethanol
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CO
1468-95-7
C15H12O
208.26
MFCD00001264
JCJNNHDZTLRSGN-UHFFFAOYSA-N
9-anthracenemethanol, 9-hydroxymethylanthracene, 9-anthrylcarbinol, 9-methylolanthracene, 9-anthrylmethanol, 9-hydroxymethylantracene, 9-anthracene methanol, 9-hydroxymethyl anthracene, anthracene-9-carbinol, ccris 7297
73848
anthracen-9-ylmethanol
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CO

An anthracene derivative used in Diels-Alder reactions. Also used in the preparation of 9-anthracenylmethyl-1-piperazinecarboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An anthracene derivative used in Diels-Alder reactions. Also used in the preparation of 9-anthracenylmethyl-1-piperazinecarboxylate.

Solubility
Soluble in hot methanol very faint turbidity.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 160°C to 165°C
Quantity 10 g
Beilstein 1873402
Solubility Information Soluble in hot methanol very faint turbidity.
Formula Weight 208.26
Percent Purity 97%
Chemical Name or Material 9-Anthracenemethanol
EINECSNumber 215-998-5
RTECSNumber CB0577500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit