Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Anthracenemethanol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
250g
Packaging:
Glass bottle
1468-95-7
C15H12O
208.26
MFCD00001264
JCJNNHDZTLRSGN-UHFFFAOYSA-N
9-anthracenemethanol,9-hydroxymethylanthracene,9-anthrylcarbinol,9-methylolanthracene,9-anthrylmethanol,9-hydroxymethylantracene,9-anthracene methanol,9-hydroxymethyl anthracene,anthracene-9-carbinol,ccris 7297
73848
anthracen-9-ylmethanol
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CO
1468-95-7
C15H12O
208.26
MFCD00001264
JCJNNHDZTLRSGN-UHFFFAOYSA-N
9-anthracenemethanol,9-hydroxymethylanthracene,9-anthrylcarbinol,9-methylolanthracene,9-anthrylmethanol,9-hydroxymethylantracene,9-anthracene methanol,9-hydroxymethyl anthracene,anthracene-9-carbinol,ccris 7297
73848
anthracen-9-ylmethanol
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CO
Packaging Glass bottle
Color Yellow
Melting Point 160.0°C to 164.0°C
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 208.26
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 9-Anthracenemethanol, 98%
Show More Show Less

EINECSNumber : 215-998-5

RTECSNumber : CB0577500

TSCA : TSCA