Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Bromo-1-nonanol, 97%, Acros Organics™

Click to view available options
Quantity:
5g
25g
55362-80-6
C9H19BrO
223.16
USJDOLXCPFASNV-UHFFFAOYSA-N
9-bromo-1-nonanol,1-nonanol, 9-bromo,9-bromo-nonan-1-ol,nonamethylene bromohydrin,9-bromnonan-1-ol,9-bromononanol,9-bromononyl alcohol,pubchem23429,9-bromo nonanol,acmc-1axfq
108700
9-bromononan-1-ol
C(CCCCO)CCCCBr
55362-80-6
C9H19BrO
223.16
USJDOLXCPFASNV-UHFFFAOYSA-N
9-bromo-1-nonanol,1-nonanol, 9-bromo,9-bromo-nonan-1-ol,nonamethylene bromohydrin,9-bromnonan-1-ol,9-bromononanol,9-bromononyl alcohol,pubchem23429,9-bromo nonanol,acmc-1axfq
108700
9-bromononan-1-ol
C(CCCCO)CCCCBr
Quantity 5g
Infrared Spectrum Authentic
Flash Point 109°C
Boiling Point 125.0°C to 126.0°C (2.0 Torr)
Melting Point 30.0°C to 33.0°C
Assay Percent Range 96% min. (GC)
Formula Weight 223.16
Percent Purity 97%
Chemical Name or Material 9-Bromo-1-nonanol

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning