Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 9-Bromo-1-nonene, 97%

Click to view available options
Quantity:
1g
5g
89359-54-6
C9H17Br
205.139
MFCD09037826
RQXPBVHYVAOUBY-UHFFFAOYSA-N
9-bromo-1-nonene,1-nonene, 9-bromo,8-nonenyl bromide,1-bromo-8-nonene,9-bromanylnon-1-ene
11019998
9-bromonon-1-ene
C=CCCCCCCCBr
89359-54-6
C9H17Br
205.139
MFCD09037826
RQXPBVHYVAOUBY-UHFFFAOYSA-N
9-bromo-1-nonene,1-nonene, 9-bromo,8-nonenyl bromide,1-bromo-8-nonene,9-bromanylnon-1-ene
11019998
9-bromonon-1-ene
C=CCCCCCCCBr
Applications
It′s employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Boiling Point 100°C to 101°C (15mmHg)
Refractive Index 1.4675
Quantity 1g
Color Colorless to Yellow
Linear Formula Br(CH2)7CH=CH2
Beilstein 1746338
Formula Weight 205.14
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 9-Bromo-1-nonene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning