Alfa Aesar™ 9-Bromofluorene, 98+%

Manufacturer: Alfa Aesar™ A1042830

Quantity 250g

 View more versions of this product

Catalog No. AAA1042830

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1940-57-4
    Molecular Formula C13H9Br
    Formula Weight 245.12
    MDL Number MFCD00001133
    UN Number UN3261
    Beilstein 2047220
    Chemical Name or Material 9-Bromofluorene

    Specifications

    Formula Weight 245.12
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Molecular Formula C13H9Br
    UN Number UN3261
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Quantity 250g
    CAS 1940-57-4
    Beilstein 2047220
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Melting Point 101° to 105°C
    Odor Irritating
    MDL Number MFCD00001133
    RTECS Number LL5890000
    Chemical Name or Material 9-Bromofluorene
    Assay ≥98%

    Description

    Applications
    9-Bromofluorene, is used to treat skin dermatological conditions and in pharmaceutical industry.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Air sensitive. Stable under normal conditions. Preserve in light-resistant and well-closed containers

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit