Alfa Aesar™ 9-Chlorofluorene, 98%

Manufacturer: Alfa Aesar™ H5035206

Quantity 5g

 View more versions of this product

Catalog No. AAH5035206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6630-65-5
    Molecular Formula C13H9Cl
    Formula Weight 200.67
    MDL Number MFCD00043468
    Chemical Name or Material 9-Chlorofluorene

    Specifications

    Formula Weight 200.67
    Molecular Formula C13H9Cl
    MDL Number MFCD00043468
    CAS 6630-65-5
    RTECS Number LL6930000
    Melting Point 86° to 88°C
    Quantity 5g
    Chemical Name or Material 9-Chlorofluorene
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit