Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-(Chloromethyl)anthracene, 98+%, ACROS Organics™

Click to view available options
Quantity:
2.5g
10g
Packaging:
Glass bottle
24463-19-2
C15H11Cl
226.7
MFCD00001263
PCVRSXXPGXRVEZ-UHFFFAOYSA-N
9-chloromethyl anthracene,anthracene, 9-chloromethyl,9-anthracenylmethyl chloride,ccris 7993,9-chloromethyanthracene,pubchem14854,9-chlormethyl-anthracen,acmc-209tez
32385
9-(chloromethyl)anthracene
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CCl
24463-19-2
C15H11Cl
226.7
MFCD00001263
PCVRSXXPGXRVEZ-UHFFFAOYSA-N
9-chloromethyl anthracene,anthracene, 9-chloromethyl,9-anthracenylmethyl chloride,ccris 7993,9-chloromethyanthracene,pubchem14854,9-chlormethyl-anthracen,acmc-209tez
32385
9-(chloromethyl)anthracene
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CCl
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 136°C to 141°C
Quantity 2.5g
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (HPLC)
Formula Weight 226.7
Physical Form Powder
Percent Purity 98+%
Chemical Name or Material 9-(Chloromethyl)anthracene, 98+%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 246-274-7

RTECSNumber : CA9600000