Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Decen-1-ol 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
13019-22-2
C10H20O
156.27
QGFSQVPRCWJZQK-UHFFFAOYSA-N
9-decen-1-ol,decylenic alcohol,1-decen-10-ol,omega-decenol,9-decenol,omega-decen-1-ol,.omega.-decen-1-ol,.omega.-decenol,9-decene-1-ol,9-decene1-ol
25612
dec-9-en-1-ol
C=CCCCCCCCCO
13019-22-2
C10H20O
156.27
QGFSQVPRCWJZQK-UHFFFAOYSA-N
9-decen-1-ol,decylenic alcohol,1-decen-10-ol,omega-decenol,9-decenol,omega-decen-1-ol,.omega.-decen-1-ol,.omega.-decenol,9-decene-1-ol,9-decene1-ol
25612
dec-9-en-1-ol
C=CCCCCCCCCO
Viscosity 11.4 mPa.s (20°C)
Density 0.8450g/mL
Flash Point 104°C
Boiling Point 234.0°C to 238.0°C
Quantity 100g
Melting Point -13.0°C
Packaging Glass bottle
Specific Gravity 0.845
Refractive Index 1.4470 to 1.4490
Infrared Spectrum Authentic
Color Orange-Red to Red
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula H2C=CH(CH2)8OH
Beilstein 01,451
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 156.27
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 9-Decen-1-ol, 99%
Show More Show Less

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 235-878-6

RTECSNumber : HE2095000

TSCA : TSCA