Alfa Aesar™ 9-Dodecyn-1-ol, 97%

Manufacturer: Alfa Aesar™ B2257706

Quantity 5g

 View more versions of this product

Catalog No. AAB2257706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 71084-08-7
    Molecular Formula C12H22O
    Linear Formula CH3CH2C≡C(CH2)8OH
    Formula Weight 182.31
    MDL Number MFCD00082940
    Beilstein 6642771
    Chemical Name or Material 9-Dodecyn-1-ol

    Specifications

    Formula Weight 182.31
    TSCA TSCA
    Molecular Formula C12H22O
    Linear Formula CH3CH2C≡C(CH2)8OH
    MDL Number MFCD00082940
    CAS 71084-08-7
    Beilstein 6642771
    Quantity 5g
    Chemical Name or Material 9-Dodecyn-1-ol
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit