Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 9-Dodecyn-1-ol, 97%

Click to view available options
Quantity:
5g
25g
71084-08-7
C12H22O
182.307
MFCD00082940
MIDXCKJJIFQOLT-UHFFFAOYSA-N
9-dodecyn-1-ol
117011
dodec-9-yn-1-ol
CCC#CCCCCCCCCO
71084-08-7
C12H22O
182.307
MFCD00082940
MIDXCKJJIFQOLT-UHFFFAOYSA-N
9-dodecyn-1-ol
117011
dodec-9-yn-1-ol
CCC#CCCCCCCCCO
Quantity 25g
Linear Formula CH3CH2C≡C(CH2)8OH
Beilstein 6642771
Formula Weight 182.31
Percent Purity 97%
Chemical Name or Material 9-Dodecyn-1-ol