Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 9-Ethylcarbazole-3-boronic acid, 98%

Click to view available options
Quantity:
250mg
1g
5g
669072-93-9
C14H14BNO2
239.081
MFCD09743121
JUBKLYCRFZTXBA-UHFFFAOYSA-N
9-ethyl-9h-carbazol-3-yl boronic acid,9-ethyl-3-carbazole boronic acid,boronic acid, 9-ethyl-9h-carbazol-3-yl,ksc924s3d,9-ethylcarbazole-3-boronic acid,9-ethyl-9h-carbazol-3-yl boronicacid
45382262
(9-ethylcarbazol-3-yl)boronic acid
B(C1=CC2=C(C=C1)N(C3=CC=CC=C32)CC)(O)O
669072-93-9
C14H14BNO2
239.081
MFCD09743121
JUBKLYCRFZTXBA-UHFFFAOYSA-N
9-ethyl-9h-carbazol-3-yl boronic acid,9-ethyl-3-carbazole boronic acid,boronic acid, 9-ethyl-9h-carbazol-3-yl,ksc924s3d,9-ethylcarbazole-3-boronic acid,9-ethyl-9h-carbazol-3-yl boronicacid
45382262
(9-ethylcarbazol-3-yl)boronic acid
B(C1=CC2=C(C=C1)N(C3=CC=CC=C32)CC)(O)O
Quantity 250mg
Beilstein 23374049
Formula Weight 239.08
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 9-Ethylcarbazole-3-boronic acid