Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Ethynyl-9-fluorenol, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass Bottle
13461-74-0
C15H10O
206.244
MFCD00041555
MMZVVJGCZZAWBN-UHFFFAOYSA-N
9-ethynyl-9-fluorenol,9-ethynyl-9h-fluoren-9-ol,acmc-209bwd,9-ethynyl-9-hydroxyfluorene,9-ethinyl-9-hydroxy-fluorene,9h-fluoren-9-ol,9-ethynyl,9-ethynyl-9h-fluoren-9-ol #,mmzvvjgczzawbn-uhfffaoysa,9-hydroxy-9-fluorenyl acetylene,9-ethynyl-9-fluorenol,
518771
9-ethynylfluoren-9-ol
C#CC1(C2=CC=CC=C2C3=CC=CC=C31)O
13461-74-0
C15H10O
206.244
MFCD00041555
MMZVVJGCZZAWBN-UHFFFAOYSA-N
9-ethynyl-9-fluorenol,9-ethynyl-9h-fluoren-9-ol,acmc-209bwd,9-ethynyl-9-hydroxyfluorene,9-ethinyl-9-hydroxy-fluorene,9h-fluoren-9-ol,9-ethynyl,9-ethynyl-9h-fluoren-9-ol #,mmzvvjgczzawbn-uhfffaoysa,9-hydroxy-9-fluorenyl acetylene,9-ethynyl-9-fluorenol,
518771
9-ethynylfluoren-9-ol
C#CC1(C2=CC=CC=C2C3=CC=CC=C31)O
Packaging Glass Bottle
Quantity 5g
Infrared Spectrum Authentic
Color White
Melting Point 107°C to 110°C
Formula Weight 206.24
Physical Form Crystalline Powder and Chunks
Percent Purity ≥98.5%
Chemical Name or Material 9-Ethynyl-9-fluorenol

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.