Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Fluorenone-2-carboxylic Acid 97%, ACROS Organics™

Click to view available options
Quantity:
1g
784-50-9
C14H8O3
224.22
MFCD00001156
BJCTXUUKONLPPK-UHFFFAOYSA-N
9-fluorenone-2-carboxylic acid,9-oxo-9h-fluorene-2-carboxylic acid,fluorenone-2-carboxylic acid,9-oxofluoren-2-carboxylic acid,9h-fluorene-2-carboxylic acid, 9-oxo,9-fluorenone-2-carboxylicacid,9-oxo-2-fluorenecarboxylic acid,bjctxuukonlppk-uhfffaoysa,9-oxo-2-fluorene carboxylic acid,9-oxo-fluorene-2-carboxylic acid
69913
9-oxofluorene-2-carboxylic acid
C1=CC=C2C(=C1)C3=C(C2=O)C=C(C=C3)C(=O)O
784-50-9
C14H8O3
224.22
MFCD00001156
BJCTXUUKONLPPK-UHFFFAOYSA-N
9-fluorenone-2-carboxylic acid,9-oxo-9h-fluorene-2-carboxylic acid,fluorenone-2-carboxylic acid,9-oxofluoren-2-carboxylic acid,9h-fluorene-2-carboxylic acid, 9-oxo,9-fluorenone-2-carboxylicacid,9-oxo-2-fluorenecarboxylic acid,bjctxuukonlppk-uhfffaoysa,9-oxo-2-fluorene carboxylic acid,9-oxo-fluorene-2-carboxylic acid
69913
9-oxofluorene-2-carboxylic acid
C1=CC=C2C(=C1)C3=C(C2=O)C=C(C=C3)C(=O)O
Melting Point >300°C
Infrared Spectrum Authentic
Quantity 1g
Color Yellow
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 10, 774
Formula Weight 224.22
Physical Form Fine Powder
Percent Purity ≥96.0%
Chemical Name or Material 9-Fluorenone-2-carboxylic acid, 97%
Show More Show Less

EINECSNumber : 212-317-3