Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Fluorenone hydrazone, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
13629-22-6
C13H10N2
194.24
YCNUILAKOMIBAL-UHFFFAOYSA-N
9-fluorenone hydrazone,fluoren-9-one, hydrazone,fluorenone hydrazone,9h-fluoren-9-ylidene hydrazine,9h-fluoren-9-one, hydrazone,fluoren-9-one hydrazone,9h-fluoren-9-ylidenehydrazine,fluorenonhydrazon,fluoren-9-ylidenediazane,fluoren-9-ylidene-hydrazine
83627
fluoren-9-ylidenehydrazine
C1=CC=C2C(=C1)C3=CC=CC=C3C2=NN
13629-22-6
C13H10N2
194.24
YCNUILAKOMIBAL-UHFFFAOYSA-N
9-fluorenone hydrazone,fluoren-9-one, hydrazone,fluorenone hydrazone,9h-fluoren-9-ylidene hydrazine,9h-fluoren-9-one, hydrazone,fluoren-9-one hydrazone,9h-fluoren-9-ylidenehydrazine,fluorenonhydrazon,fluoren-9-ylidenediazane,fluoren-9-ylidene-hydrazine
83627
fluoren-9-ylidenehydrazine
C1=CC=C2C(=C1)C3=CC=CC=C3C2=NN
Packaging Glass bottle
Quantity 25g
Melting Point 148.0°C to 150.0°C
Assay Percent Range 97%
Beilstein 07,I,251
Solubility Information Solubility in water: insoluble
Formula Weight 194.24
Percent Purity 97%
Chemical Name or Material 9-Fluorenone hydrazone