Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 9-Fluorenone hydrazone, 98+%

Click to view available options
Quantity:
5g
25g
13629-22-6
C13H10N2
194.237
MFCD00016357
YCNUILAKOMIBAL-UHFFFAOYSA-N
9-fluorenone hydrazone,fluoren-9-one, hydrazone,fluorenone hydrazone,9h-fluoren-9-ylidene hydrazine,9h-fluoren-9-one, hydrazone,fluoren-9-one hydrazone,9h-fluoren-9-ylidenehydrazine,fluorenonhydrazon,fluoren-9-ylidenediazane,fluoren-9-ylidene-hydrazine
83627
fluoren-9-ylidenehydrazine
C1=CC=C2C(=C1)C3=CC=CC=C3C2=NN
13629-22-6
C13H10N2
194.237
MFCD00016357
YCNUILAKOMIBAL-UHFFFAOYSA-N
9-fluorenone hydrazone,fluoren-9-one, hydrazone,fluorenone hydrazone,9h-fluoren-9-ylidene hydrazine,9h-fluoren-9-one, hydrazone,fluoren-9-one hydrazone,9h-fluoren-9-ylidenehydrazine,fluorenonhydrazon,fluoren-9-ylidenediazane,fluoren-9-ylidene-hydrazine
83627
fluoren-9-ylidenehydrazine
C1=CC=C2C(=C1)C3=CC=CC=C3C2=NN
Quantity 5g
Melting Point 150°C to 154°C
Beilstein 1910575
Formula Weight 194.24
Percent Purity ≥98%
Chemical Name or Material 9-Fluorenone hydrazone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning