Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 9-Fluorenylmethyl pentafluorophenyl carbonate, 98%

Click to view available options
Quantity:
1g
5g
25g
88744-04-1
C21H11F5O3
406.308
MFCD00013261
CBBKZVZOEBSFQX-UHFFFAOYSA-N
9-fluorenylmethyl pentafluorophenyl carbonate,fmoc-opfp,9h-fluoren-9-yl methyl perfluorophenyl carbonate,9h-fluoren-9-ylmethyl 2,3,4,5,6-pentafluorophenyl carbonate,9h-fluoren-9-ylmethyl pentafluorophenyl carbonate,carbonic acid 9h-fluoren-9-ylmethyl pentafluorophenyl ester,carbonic acid,9h-fluoren-9-ylmethyl 2,3,4,5,6-pentafluorophenyl ester,acmc-20ecck,fmoc pentafluorophenyl ester,9-fluorenylmethylpentafluorophenylcarbonate
2733201
9H-fluoren-9-ylmethyl (2,3,4,5,6-pentafluorophenyl) carbonate
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)OC4=C(C(=C(C(=C4F)F)F)F)F
88744-04-1
C21H11F5O3
406.308
MFCD00013261
CBBKZVZOEBSFQX-UHFFFAOYSA-N
9-fluorenylmethyl pentafluorophenyl carbonate,fmoc-opfp,9h-fluoren-9-yl methyl perfluorophenyl carbonate,9h-fluoren-9-ylmethyl 2,3,4,5,6-pentafluorophenyl carbonate,9h-fluoren-9-ylmethyl pentafluorophenyl carbonate,carbonic acid 9h-fluoren-9-ylmethyl pentafluorophenyl ester,carbonic acid,9h-fluoren-9-ylmethyl 2,3,4,5,6-pentafluorophenyl ester,acmc-20ecck,fmoc pentafluorophenyl ester,9-fluorenylmethylpentafluorophenylcarbonate
2733201
9H-fluoren-9-ylmethyl (2,3,4,5,6-pentafluorophenyl) carbonate
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)OC4=C(C(=C(C(=C4F)F)F)F)F
Melting Point 83°C to 86°C
Quantity 5g
Formula Weight 406.31
Percent Purity 98%
Chemical Name or Material 9-Fluorenylmethyl pentafluorophenyl carbonate

Recommended Storage : 2° to 8°C