Alfa Aesar™ 9-Fluorenylmethyl pentafluorophenyl carbonate, 98%

Manufacturer: Alfa Aesar™ B2229703

Quantity 1g

 View more versions of this product

Catalog No. AAB2229703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 88744-04-1
    Molecular Formula C21H11F5O3
    Formula Weight 406.31
    MDL Number MFCD00013261
    Chemical Name or Material 9-Fluorenylmethyl pentafluorophenyl carbonate

    Specifications

    Formula Weight 406.31
    Molecular Formula C21H11F5O3
    MDL Number MFCD00013261
    CAS 88744-04-1
    Recommended Storage 2° to 8°C
    Melting Point 83° to 86°C
    Quantity 1g
    Chemical Name or Material 9-Fluorenylmethyl pentafluorophenyl carbonate
    Assay 98%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit