Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Fluorenylmethyl 1-benzotriazolyl carbonate, 98%, Thermo Scientific™

Catalog Number ac441510050
Click to view available options
Quantity:
5 g
82911-71-5
C21H15N3O3
357.37
KNSPZTVPSAKHBN-UHFFFAOYSA-N
fmoc-obt, 9h-fluoren-9-yl methyl 1h-benzo d 1,2,3 triazol-1-yl carbonate, 9-fluorenylmethyl 1-benzotriazolyl carbonate, benzotriazol-1-yl 9-fluorenylmethyl carbonate, 1,2,3-benzotriazol-1-yl 9h-fluoren-9-ylmethyl carbonate, 1-fmoc-oxy benzotriazole, 1-fmoc-oxy-1h-benzotriazole, 9-fluorenylmethyl benzotriazolycarbonate, 9-fluorenylmethylbenzotriazolylcarbonate, 9-fluorenylmethyl 1-benzotriazolyl &
5236567
benzotriazol-1-yl 9H-fluoren-9-ylmethyl carbonate
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)ON4C5=CC=CC=C5N=N4
82911-71-5
C21H15N3O3
357.37
KNSPZTVPSAKHBN-UHFFFAOYSA-N
fmoc-obt, 9h-fluoren-9-yl methyl 1h-benzo d 1,2,3 triazol-1-yl carbonate, 9-fluorenylmethyl 1-benzotriazolyl carbonate, benzotriazol-1-yl 9-fluorenylmethyl carbonate, 1,2,3-benzotriazol-1-yl 9h-fluoren-9-ylmethyl carbonate, 1-fmoc-oxy benzotriazole, 1-fmoc-oxy-1h-benzotriazole, 9-fluorenylmethyl benzotriazolycarbonate, 9-fluorenylmethylbenzotriazolylcarbonate, 9-fluorenylmethyl 1-benzotriazolyl &
5236567
benzotriazol-1-yl 9H-fluoren-9-ylmethyl carbonate
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)ON4C5=CC=CC=C5N=N4
CAS Min % 97.5
CAS Max % 100.0
Melting Point 165°C to 167°C
Color White
Elemental Analysis 68.82 to 72.34% C, 11.46 to 12.05% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 5 g
Formula Weight 357.37
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 9-Fluorenylmethyl 1-benzotriazolyl carbonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit