Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Hydroxy-4-methoxyacridine, 97%

Click to view available options
Quantity:
1g
35308-00-0
C14H11NO2
225.247
MFCD00005027
ZYEVJRPVZZGDRM-UHFFFAOYSA-N
4-methoxyacridin-9-ol,9-hydroxy-4-methoxyacridine,4-methoxy-9-acridinol,4-methoxyacridin-9 10h-one,9-acridinol, 4-methoxy,10h-acridin-9-one, 4-methoxy,9 10h-acridinone, 4-methoxy,4-methoxy-9-acridanone,4-methoxyacridon,acmc-20ah2d
51991
4-methoxy-10H-acridin-9-one
COC1=CC=CC2=C1NC3=CC=CC=C3C2=O
35308-00-0
C14H11NO2
225.247
MFCD00005027
ZYEVJRPVZZGDRM-UHFFFAOYSA-N
4-methoxyacridin-9-ol,9-hydroxy-4-methoxyacridine,4-methoxy-9-acridinol,4-methoxyacridin-9 10h-one,9-acridinol, 4-methoxy,10h-acridin-9-one, 4-methoxy,9 10h-acridinone, 4-methoxy,4-methoxy-9-acridanone,4-methoxyacridon,acmc-20ah2d
51991
4-methoxy-10H-acridin-9-one
COC1=CC=CC2=C1NC3=CC=CC=C3C2=O
Quantity 1g
Melting Point ~286°C
Beilstein 200297
Formula Weight 225.25
Percent Purity 97%
Chemical Name or Material 9-Hydroxy-4-methoxyacridine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning