Alfa Aesar™ 9-Hydroxy-9-fluorenecarboxylic acid, 97%

Manufacturer: Alfa Aesar™ B2240906

Quantity 5g

 View more versions of this product

Catalog No. AAB2240906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 467-69-6
    Molecular Formula C14H10O3
    Formula Weight 226.23
    MDL Number MFCD00001137
    UN Number UN3077
    Beilstein 1314711
    Chemical Name or Material 9-Hydroxy-9-fluorenecarboxylic acid

    Specifications

    Formula Weight 226.23
    TSCA TSCA
    MDL Number MFCD00001137
    CAS 467-69-6
    UN Number UN3077
    Beilstein 1314711
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Melting Point ≈164°C (dec.)
    Quantity 5g
    Molecular Formula C14H10O3
    RTECS Number LL6480000
    Chemical Name or Material 9-Hydroxy-9-fluorenecarboxylic acid
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit