Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 9-Hydroxy-9-fluorenecarboxylic acid, 97%

Click to view available options
Quantity:
5g
25g
100g
467-69-6
C14H10O3
226.231
MFCD00001137
GXAMYUGOODKVRM-UHFFFAOYSA-N
flurenol,9-hydroxy-9-fluorenecarboxylic acid,flurecol,9-hydroxy-9h-fluorene-9-carboxylic acid,glycolic acid, diphenylene,9h-fluorene-9-carboxylic acid, 9-hydroxy,flurecol bsi:iso,unii-4tkt65am8p,9-hydooxyfluorene-9-carboxylic acid,9-hydroxy-9-fluorene carboxyli acid
10087
CHEBI:82015
9-hydroxyfluorene-9-carboxylic acid
C1=CC=C2C(=C1)C3=CC=CC=C3C2(C(=O)O)O
467-69-6
C14H10O3
226.231
MFCD00001137
GXAMYUGOODKVRM-UHFFFAOYSA-N
flurenol,9-hydroxy-9-fluorenecarboxylic acid,flurecol,9-hydroxy-9h-fluorene-9-carboxylic acid,glycolic acid, diphenylene,9h-fluorene-9-carboxylic acid, 9-hydroxy,flurecol bsi:iso,unii-4tkt65am8p,9-hydooxyfluorene-9-carboxylic acid,9-hydroxy-9-fluorene carboxyli acid
10087
CHEBI:82015
9-hydroxyfluorene-9-carboxylic acid
C1=CC=C2C(=C1)C3=CC=CC=C3C2(C(=O)O)O
Melting Point ~164°C (decomposition)
Quantity 25g
UN Number UN3077
Beilstein 1314711
Formula Weight 226.23
Percent Purity 97%
Chemical Name or Material 9-Hydroxy-9-fluorenecarboxylic acid

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : LL6480000

TSCA : TSCA