Alfa Aesar™ 9-Methylcarbazole-3-boronic acid pinacol ester, 95%

Manufacturer: Alfa Aesar™ H5195206

Quantity 5g

 View more versions of this product

Catalog No. AAH5195206

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C19H22BNO2
    Formula Weight 307.2
    MDL Number MFCD16294552
    Synonym 9-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
    Chemical Name or Material 9-Methylcarbazole-3-boronic acid pinacol ester

    Specifications

    Formula Weight 307.2
    Molecular Formula C19H22BNO2
    MDL Number MFCD16294552
    Synonym 9-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
    Quantity 5g
    Chemical Name or Material 9-Methylcarbazole-3-boronic acid pinacol ester
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit